siRNA Details
Gene Symbol: Creb3l4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB047647.1 | - | 1 | 168 | ||
AB052779.1 | - | 3 | 425 | |||
AB182651.1 | - | 3 | 543 | |||
AB182652.1 | - | 1 | 168 | |||
AF287260.1 | - | 1 | 306 | |||
AK005918.1 | - | 1 | 325 | |||
AK019928.1 | - | 3 | 543 | |||
AK088919.1 | - | 3 | 442 | |||
AY049978.1 | - | 3 | 464 | |||
BC022605.1 | - | 3 | 399 | 4164994 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
