siRNA Details
Gene Symbol: Nav2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111016.1 | NP_001104486.1 | 4, 5 | 912 | ||
NM_175272.3 | NP_780481.2 | 3, 4 | 1174 | |||
GenBank mRNA | AK031334.1 | - | 3, 4 | 472 | ||
AK039374.1 | - | 3, 4 | 1182 | |||
AK040152.1 | - | 2, 3 | 459 | |||
AK084063.1 | - | 4, 5 | 890 | |||
AK129480.1 | - | 2, 3 | 400 | |||
AK158337.1 | - | 3, 4 | 586 | |||
BC150679.1 | - | 2, 3 | 162 |
