siRNA Details
Gene Symbol: Naa38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030083.2 | NP_084359.1 | 2 | 242 | ||
GenBank mRNA | AK018782.1 | - | 2 | 243 | ||
BC038659.1 | - | 2 | 196 | |||
BC048499.1 | - | 2 | 248 | 6583055 | ||
BC062267.1 | - | 2 | 212 | 1532489 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
