siRNA Details
Gene Symbol: Fam219b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166364.1 | NP_001159836.1 | 2, 3 | 399 | ||
NM_175273.4 | NP_780482.1 | 2, 3 | 399 | |||
GenBank mRNA | AK028365.1 | - | 2, 3 | 400 | ||
AK185896.1 | - | 1, 2 | 60 | |||
BC111883.1 | - | 2, 3 | 364 | 40060305 | ||
BC112403.1 | - | 2, 3 | 364 | 40060304 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001108764, XM_008766309, XM_008766310.
Gene Ontology Categories:
