siRNA Details
Gene Symbol: 2210407C18Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144544.2 | NP_653127.2 | 5 | 481 | ||
GenBank mRNA | AA124355.1 | - | 5 | 467 | ||
AA434919.1 | - | 3 | 287 | |||
AY148487.1 | - | 4 | 387 | |||
BC019553.1 | - | 5 | 459 | 4456658 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
