siRNA Details
Gene Symbol: Tmem247 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277980.1 | NP_001264909.1 | 3 | 732 | ||
NM_030104.1 | NP_084380.1 | 3 | 729 | |||
GenBank mRNA | AK018916.1 | - | 3 | 663 | ||
AK076910.1 | - | 3 | 640 | |||
BC100486.1 | - | 3 | 643 |
