siRNA Details
Gene Symbol: Arhgap10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030113.2 | NP_084389.2 | 7, 8 | 831 | ||
GenBank mRNA | AF297029.1 | - | 7, 8 | 735 | ||
AF297030.1 | - | 7, 8 | 794 | |||
AY179965.1 | - | 7, 8 | 707 | |||
AY179966.1 | - | 7, 8 | 707 | |||
AY179967.1 | - | 7, 8 | 707 | |||
CN703944.1 | - | 18 | ||||
CV783737.1 | - | 2, 3 | 209 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001109501, XM_008772532, XM_008772531, XM_008772530.
Gene Ontology Categories:
