siRNA Details
Gene Symbol: 1700061G19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030141.1 | NP_084417.1 | 14 | 2286 | ||
GenBank mRNA | AK028078.1 | - | 14 | 2437 | ||
BC116283.1 | - | 14 | 2286 | 40094427 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
