siRNA Details
Gene Symbol: Spaca7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024279.2 | NP_077241.1 | 3 | 289 | ||
GenBank mRNA | AK018926.1 | - | 3 | 290 | ||
BC048522.1 | - | 3 | 289 | 6741699 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
