siRNA Details
Gene Symbol: Lsm6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191004.1 | NP_001177933.1 | 4 | 362 | ||
NM_030145.3 | NP_084421.1 | 3 | 263 | |||
GenBank mRNA | AK005329.1 | - | 1 | 204 | ||
AK019126.1 | - | 4 | 322 | |||
AK078392.1 | - | 4 | 312 | |||
AK141884.1 | - | 4 | 309 | |||
AK158236.1 | - | 3 | 252 | |||
AK161686.1 | - | 4 | 316 | |||
BC030427.1 | - | 3 | 210 | 5370660 | ||
BC098230.1 | - | 3 | 213 | 3372313 | ||
BC099484.1 | - | 3 | 268 | 30936207 | ||
DV648841.1 | - | 4 | 362 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
