siRNA Details
Gene Symbol: Bola3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175277.4 | NP_780486.1 | 3 | 273 | ||
GenBank mRNA | AK019037.1 | - | 2 | 108 | ||
AK028102.1 | - | 3 | 274 | |||
AK131950.1 | - | 282 | ||||
AK160488.1 | - | 3 | 277 | |||
AK161878.1 | - | 45 | ||||
AK218122.1 | - | 1 | 90 | |||
BC049748.1 | - | 3 | 293 | |||
BC116947.1 | - | 4 | 311 | 40126266 | ||
BC116949.1 | - | 4 | 311 | 40126268 |

More Information
Other Species Targeted

Gene Ontology Categories:
