siRNA Details
Gene Symbol: Bola3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175277.4 | NP_780486.1 | 3 | 220 | ||
GenBank mRNA | AK019037.1 | - | 2 | 55 | ||
AK028102.1 | - | 3 | 221 | |||
AK131950.1 | - | 229 | ||||
AK160488.1 | - | 3 | 224 | |||
AK218122.1 | - | 1 | 37 | |||
BC049748.1 | - | 3 | 240 | |||
BC116947.1 | - | 4 | 258 | 40126266 | ||
BC116949.1 | - | 4 | 258 | 40126268 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
