siRNA Details
Gene Symbol: Brd8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289606.1 | NP_001276535.1 | 13 | 2081 | ||
NM_001289607.1 | NP_001276536.1 | 13 | 2090 | |||
NM_030147.3 | NP_084423.2 | 14 | 2300 | |||
GenBank mRNA | AK032320.1 | - | 14 | 2012 | ||
AK132365.1 | - | 14 | 1996 | |||
AK161689.1 | - | 13 | 1794 | |||
BC021482.1 | - | 13 | 3176 | |||
BC023837.1 | - | 13 | 1806 | |||
BC025644.1 | - | 14 | 2045 | 5323755 |
