siRNA Details
Gene Symbol: Brd8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289606.1 | NP_001276535.1 | 11 | 1397 | ||
NM_001289607.1 | NP_001276536.1 | 11 | 1406 | |||
NM_030147.3 | NP_084423.2 | 12 | 1616 | |||
GenBank mRNA | AK032320.1 | - | 12 | 1328 | ||
AK132365.1 | - | 12 | 1312 | |||
AK161689.1 | - | 11 | 1110 | |||
BC021482.1 | - | 11 | 2492 | |||
BC023837.1 | - | 11 | 1122 | |||
BC025644.1 | - | 12 | 1361 | 5323755 |

More Information
Other Species Targeted

Gene Ontology Categories:
