siRNA Details
Gene Symbol: D730001G18Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK021322.1 | - | 2 | 429 | ||
AK037167.1 | - | 2 | 428 | |||
BC055822.1 | - | 3 | 492 | |||
BC147771.1 | - | 1 | 308 | |||
BC147774.1 | - | 1 | 308 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
