siRNA Details
Gene Symbol: Rassf10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175279.3 | NP_780488.2 | 1 | 896 | ||
GenBank mRNA | AK019489.1 | - | 1 | 343 | ||
AK031660.1 | - | 1 | 897 | |||
AK046433.1 | - | 1 | 892 | |||
BC028854.1 | - | 1 | 472 | |||
BC125291.1 | - | 1 | 712 | 40129806 | ||
BC125293.1 | - | 1 | 712 | 40129808 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
