siRNA Details
Gene Symbol: Fam122b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166365.2 | NP_001159837.2 | 8 | 1435 | ||
NM_001166583.1 | NP_001160055.1 | 8 | 1435 | |||
NM_030167.4 | NP_084443.1 | 8 | 1438 | |||
GenBank mRNA | AB541013.1 | - | 8 | 603 | ||
AK019480.1 | - | 8 | 665 | |||
AK036417.1 | - | 8 | 1432 | |||
AK044896.1 | - | 8 | 1436 | |||
BC066167.1 | - | 8 | 650 | 30433483 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
