siRNA Details
Gene Symbol: Mctp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030174.2 | NP_084450.2 | 15 | 1817 | ||
GenBank mRNA | AK013379.1 | - | 2 | 423 | ||
AK047562.1 | - | 15 | 1818 | |||
AK054478.1 | - | 11 | 1190 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: XM_008760611, XM_008760612, XM_008760610, XM_008760614, XM_008760616, and others.
Gene Ontology Categories:
