siRNA Details
Gene Symbol: Zc3hav1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028421.1 | NP_082697.1 | 8 | 2630 | ||
NM_028864.2 | NP_083140.1 | 8 | 2630 | |||
GenBank mRNA | AK004770.1 | - | 8 | 2631 | ||
AK020263.2 | - | 8 | 2309 | |||
AK080334.1 | - | 8 | 2615 | |||
AK143568.1 | - | 8 | 2300 | |||
AK169402.1 | - | 8 | 2275 | |||
AK172379.1 | - | 8 | 2631 | |||
BC029090.1 | - | 8 | 2192 | 2651558 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
