siRNA Details
Gene Symbol: Celf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289613.1 | NP_001276542.1 | 4 | 854 | ||
NM_001289616.1 | NP_001276545.1 | 5 | 336 | |||
NM_001289620.1 | NP_001276549.1 | 5 | 336 | |||
NM_172434.3 | NP_766022.1 | 4 | 851 | |||
GenBank mRNA | AK019578.1 | - | 2 | 563 | ||
AK030259.1 | - | 3 | 162 | |||
AK044069.1 | - | 4 | 284 | |||
AK044506.1 | - | 5 | 300 | |||
AK087536.1 | - | 5 | 296 | |||
AY165052.1 | - | 4 | 569 | |||
BC031740.1 | - | 1 | 15 | |||
BC057553.1 | - | 4 | 909 | 6401356 | ||
BY089288.1 | - | 5 | 340 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
