siRNA Details
Gene Symbol: Celf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289613.1 | NP_001276542.1 | 1 | 638 | ||
NM_001289616.1 | NP_001276545.1 | 2 | 123 | |||
NM_001289620.1 | NP_001276549.1 | 2 | 123 | |||
NM_172434.3 | NP_766022.1 | 1 | 638 | |||
GenBank mRNA | AK044069.1 | - | 1 | 71 | ||
AK044506.1 | - | 2 | 87 | |||
AK087536.1 | - | 2 | 83 | |||
AY165052.1 | - | 1 | 356 | |||
BC057553.1 | - | 1 | 693 | 6401356 | ||
BY089288.1 | - | 2 | 124 |

More Information
Other Species Targeted

Gene Ontology Categories:
