siRNA Details
Gene Symbol: Clip4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271483.1 | NP_001258412.1 | 5 | 631 | ||
NM_001271484.1 | NP_001258413.1 | 5 | 716 | |||
NM_030179.3 | NP_084455.2 | 5 | 619 | |||
NM_175378.1 | NP_780587.1 | 5 | 716 | |||
GenBank mRNA | AK017914.1 | - | 2 | 206 | ||
AK018895.1 | - | 5 | 709 | |||
AK019516.1 | - | 5 | 632 | |||
AK029962.1 | - | 5 | 717 | |||
AK075758.1 | - | 5 | 707 | |||
AK161301.1 | - | 5 | 706 | |||
BC035226.1 | - | 5 | 619 | 4920788 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
