siRNA Details
Gene Symbol: Usp54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030180.2 | NP_084456.2 | 18 | 4011 | ||
GenBank mRNA | AK019588.1 | - | 1 | 198 | ||
BC066166.1 | - | 1 | 522 | |||
BC066175.1 | - | 1 | 522 | |||
BC117844.1 | - | 1 | 186 | |||
BC117845.1 | - | 1 | 186 |
