siRNA Details
Gene Symbol: Usp54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030180.2 | NP_084456.2 | 17 | 3126 | ||
GenBank mRNA | CK638913.1 | - | 4 | 479 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001008863, XM_008770560, CB726956.
Gene Ontology Categories:
