siRNA Details
Gene Symbol: Armc9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027456.1 | NP_081732.1 | 6 | 652 | ||
NM_030184.2 | NP_084460.1 | 6 | 687 | |||
GenBank mRNA | AK014450.1 | - | 6 | 657 | ||
AK017564.1 | - | 6 | 653 | |||
AK019512.1 | - | 6 | 682 | |||
AK019600.1 | - | 6 | 676 | |||
AK030307.1 | - | 6 | 684 | |||
AK036083.1 | - | 6 | 658 | |||
AK039800.1 | - | 6 | 660 | |||
AK045699.1 | - | 6 | 651 | |||
AK046693.1 | - | 6 | 670 | |||
AK048064.1 | - | 6 | 666 | |||
AK140598.1 | - | 6 | 683 | |||
AK161393.1 | - | 6 | 667 | |||
BC043101.1 | - | 6 | 639 | 6406321 |
