siRNA Details
Gene Symbol: Ndor1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001082476.2 | NP_001075945.1 | 2, 3 | 284 | ||
NM_001252541.1 | NP_001239470.1 | 2, 3 | 284 | |||
NM_001252542.1 | NP_001239471.1 | 2, 3 | 351 | |||
GenBank mRNA | AK019617.1 | - | 2, 3 | 281 | ||
AK143124.1 | - | 2, 3 | 324 | |||
AK163207.1 | - | 2, 3 | 254 | |||
AK169885.1 | - | 2, 3 | 343 | |||
AK186395.1 | - | 67 | ||||
AK211498.1 | - | 2, 3 | 202 | |||
BC049789.1 | - | 2, 3 | 334 | 6331033 | ||
BQ771207.1 | - | 2, 3 | 251 | |||
CD354207.1 | - | 675 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
