siRNA Details
Gene Symbol: Ttc30a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030188.3 | NP_084464.3 | 1 | 1044 | ||
GenBank mRNA | AK019670.1 | - | 1 | 918 | ||
AK029535.1 | - | 847 | ||||
AK045096.1 | - | 1 | 844 | |||
BC004773.1 | - | 1 | 839 | 3256753 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
