siRNA Details
Gene Symbol: Stxbp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081344.2 | NP_001074813.2 | 25, 26 | 3666 | ||
GenBank mRNA | AF516607.1 | - | 21, 22 | 2745 | ||
AK019788.1 | - | 8, 9 | 1407 | |||
AK029982.1 | - | 6, 7 | 3417 | |||
AK122417.1 | - | 4, 5 | 745 | |||
BC038042.1 | - | 5, 6 | 1034 | |||
BC094582.1 | - | 18, 19 | 2491 |

More Information
Other Species Targeted

Gene Ontology Categories:
