siRNA Details
Gene Symbol: Stxbp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081344.2 | NP_001074813.2 | 24 | 3427 | ||
GenBank mRNA | AF516607.1 | - | 20 | 2506 | ||
AK019788.1 | - | 7 | 1168 | |||
AK029982.1 | - | 5 | 3178 | |||
AK122417.1 | - | 3 | 506 | |||
AK136998.1 | - | 3 | 1441 | |||
BC038042.1 | - | 4 | 795 | |||
BC094582.1 | - | 17 | 2252 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
