siRNA Details
Gene Symbol: Zfp623 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030199.3 | NP_084475.1 | 2 | 1514 | ||
GenBank mRNA | AK019163.1 | - | 2 | 1411 | ||
AK030476.1 | - | 2 | 1419 | |||
AK046806.1 | - | 2 | 1515 | |||
AK169386.1 | - | 2 | 1512 | |||
AK172985.1 | - | 2 | 1489 | |||
AY610936.1 | - | 2 | 1515 | |||
BC021834.1 | - | 2 | 1417 | 4022529 | ||
BC086767.1 | - | 2 | 1405 | 6832277 |
