siRNA Details
Gene Symbol: 1500015O10Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024283.3 | NP_077245.2 | 2, 3 | 414 | ||
GenBank mRNA | AK018770.1 | - | 2, 3 | 169 | ||
AK049805.1 | - | 2, 3 | 415 | |||
BC002254.1 | - | 2, 3 | 347 | 3493553 |
