siRNA Details
Gene Symbol: 1500015O10Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024283.3 | NP_077245.2 | 3 | 432 | ||
GenBank mRNA | AK018770.1 | - | 3 | 187 | ||
AK049805.1 | - | 3 | 433 | |||
BC002254.1 | - | 3 | 365 | 3493553 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
