siRNA Details
Gene Symbol: Asb15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080847.3 | NP_543123.2 | 7, 8 | 1004 | ||
GenBank mRNA | AF403043.1 | - | 7, 8 | 1005 | ||
AK019556.1 | - | 7, 8 | 1005 | |||
AK029759.1 | - | 7, 8 | 936 | |||
AK079418.1 | - | 7, 8 | 1110 | |||
BC050794.1 | - | 7, 8 | 953 | 6742480 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
