siRNA Details
Gene Symbol: Saal1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030233.1 | NP_084509.1 | 6 | 566 | ||
GenBank mRNA | AB489137.1 | - | 6 | 547 | ||
AK019878.1 | - | 6 | 567 | |||
AK030833.1 | - | 6 | 572 | |||
AK031128.1 | - | 5 | 352 | |||
AK033202.1 | - | 6 | 577 | |||
AK035874.1 | - | 6 | 553 | |||
AK132193.1 | - | 6 | 567 | |||
AK134418.1 | - | 6 | 574 | |||
BC019985.1 | - | 6 | 535 | |||
BC068127.1 | - | 6 | 563 | |||
BC087944.1 | - | 6 | 578 | 30314036 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
