siRNA Details
Gene Symbol: Saal1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030233.1 | NP_084509.1 | 10 | 1091 | ||
GenBank mRNA | AB489137.1 | - | 10 | 1072 | ||
AK019878.1 | - | 10 | 1092 | |||
AK030833.1 | - | 10 | 1097 | |||
AK031128.1 | - | 8 | 2939 | |||
AK033202.1 | - | 10 | 1102 | |||
AK035874.1 | - | 9 | 3135 | |||
AK132193.1 | - | 10 | 1092 | |||
AK134418.1 | - | 10 | 1099 | |||
BC019985.1 | - | 11 | 1441 | |||
BC028473.1 | - | 5 | 505 | |||
BC068127.1 | - | 10 | 1088 | |||
BC087944.1 | - | 10 | 1103 | 30314036 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
