siRNA Details
Gene Symbol: Fbxo34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146085.1 | NP_001139557.1 | 4 | 529 | ||
NM_001146086.1 | NP_001139558.1 | 3 | 452 | |||
NM_030236.3 | NP_084512.2 | 2 | 342 | |||
GenBank mRNA | AK013711.1 | - | 6 | 891 | ||
AK020018.1 | - | 2 | 343 | |||
AK171630.1 | - | 7 | 877 | |||
BC048149.1 | - | 4 | 563 | 5706344 | ||
BC049155.1 | - | 3 | 394 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
