siRNA Details
Gene Symbol: Popdc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024286.1 | NP_077248.1 | 4 | 1240 | ||
GenBank mRNA | AK039691.1 | - | 4 | 1092 | ||
AK052325.1 | - | 4 | 1240 | |||
BC114993.1 | - | 4 | 1063 | 40056724 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
