siRNA Details
Gene Symbol: Bhlhe41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271768.1 | NP_001258697.1 | 3 | 303 | ||
NM_024469.2 | NP_077789.1 | 3 | 303 | |||
GenBank mRNA | AA013582.1 | - | 3 | 304 | ||
AB044090.1 | - | 3 | 223 | |||
BC023231.1 | - | 3 | 255 | |||
BC037742.1 | - | 3 | 263 | |||
BC051555.1 | - | 3 | 264 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: XM_002729454, AF009329.
Gene Ontology Categories:
