siRNA Details
Gene Symbol: Sh3bp5l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK086088.1 | - | 7 | 1473 | ||
AK122543.1 | - | 5 | 4677 | |||
AK144928.1 | - | 7 | 1472 | |||
AK150129.1 | - | 7 | 1452 | |||
AK156171.1 | - | 7 | 1513 | |||
AK159202.1 | - | 7 | 1504 | |||
AK166491.1 | - | 7 | 1473 | |||
AK167203.1 | - | 7 | 1589 | |||
AK172333.1 | - | 7 | 1513 | |||
AK215492.1 | - | 1 | 65 | |||
BC003251.1 | - | 6 | 1134 | 3500047 | ||
BC013661.1 | - | 3 | 605 | |||
BC094418.1 | - | 7 | 1495 | 30702568 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
