siRNA Details
Gene Symbol: Gpr146 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001038703.2 | NP_001033792.1 | 2 | 1057 | ||
NM_030258.4 | NP_084534.2 | 2 | 982 | |||
GenBank mRNA | AK046512.1 | - | 2 | 1005 | ||
AK050034.1 | - | 2 | 951 | |||
AK087100.1 | - | 2 | 951 | |||
AK089232.1 | - | 2 | 1005 | |||
AK137454.1 | - | 2 | 1059 | |||
AK154478.1 | - | 2 | 1011 | |||
AK154708.1 | - | 2 | 1003 | |||
AK155212.1 | - | 2 | 950 | |||
AK155280.1 | - | 2 | 1071 | |||
AK155876.1 | - | 2 | 985 | |||
BC003323.1 | - | 2 | 953 | 3156060 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
