siRNA Details
Gene Symbol: Rilpl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030259.1 | NP_084535.1 | 1 | 322 | ||
GenBank mRNA | AK133760.1 | - | 1 | 454 | ||
AK150123.1 | - | 1 | 426 | |||
AK150252.1 | - | 1 | 425 | |||
AK171419.1 | - | 1 | 400 | |||
BC003324.1 | - | 1 | 322 | 3156139 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
