siRNA Details
Gene Symbol: Tusc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019742.4 | NP_062716.1 | 2 | 334 | ||
GenBank mRNA | AF123387.1 | - | 2 | 221 | ||
AK004430.1 | - | 2 | 294 | |||
AK082684.1 | - | 2 | 335 | |||
BC048477.1 | - | 2 | 334 | 6772171 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
