siRNA Details
Gene Symbol: Med8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290688.1 | NP_001277617.1 | 3 | 185 | ||
NM_020000.3 | NP_064384.2 | 3 | 225 | |||
NM_173719.3 | NP_776067.1 | 4 | 273 | |||
GenBank mRNA | AB041805.1 | - | 2 | 1711 | ||
AK005422.1 | - | 4 | 256 | |||
AK008759.1 | - | 3 | 204 | |||
AK011080.1 | - | 3 | 185 | |||
AK147032.1 | - | 3 | 211 | |||
AK148025.1 | - | 3 | 206 | |||
BC002315.1 | - | 2 | 165 | |||
BC021870.1 | - | 3 | 196 | 4022630 | ||
BC051683.1 | - | 3 | 163 | |||
BC053738.1 | - | 1 | 1072 |
