siRNA Details
Gene Symbol: Med8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290688.1 | NP_001277617.1 | 3, 4 | 276 | ||
NM_020000.3 | NP_064384.2 | 3, 4 | 316 | |||
NM_173719.3 | NP_776067.1 | 4, 5 | 364 | |||
GenBank mRNA | AB041805.1 | - | 2, 3 | 1802 | ||
AK005422.1 | - | 4, 5 | 347 | |||
AK008759.1 | - | 3, 4 | 295 | |||
AK011080.1 | - | 3, 4 | 276 | |||
AK147032.1 | - | 3, 4 | 302 | |||
AK148025.1 | - | 3, 4 | 297 | |||
BC002315.1 | - | 2, 3 | 256 | |||
BC021870.1 | - | 3, 4 | 287 | 4022630 | ||
BC051683.1 | - | 3, 4 | 254 | |||
BC053738.1 | - | 1, 2 | 1163 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
