siRNA Details
Gene Symbol: Olfr160 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030553.2 | NP_085030.2 | 1 | 638 | ||
GenBank mRNA | BC130240.1 | - | 1 | 639 | 40142140 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
