siRNA Details
Gene Symbol: Rab27b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB067775.1 | - | 4 | 577 | ||
AB120512.1 | - | 3 | 321 | |||
AB232623.1 | - | 3 | 321 | |||
AF328893.1 | - | 4 | 590 | |||
AK049854.1 | - | 5 | 474 | |||
AY010898.1 | - | 3 | 321 | |||
BC018443.1 | - | 4 | 557 | 3601865 | ||
BC032928.1 | - | 4 | 510 | 5120275 | ||
BC096496.1 | - | 5 | 986 | 30738460 | ||
BY721478.1 | - | 5 | 472 |
