siRNA Details
Gene Symbol: Rab27b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB067775.1 | - | 2 | 332 | ||
AB120512.1 | - | 1 | 76 | |||
AB232623.1 | - | 1 | 76 | |||
AF328893.1 | - | 2 | 345 | |||
AK049854.1 | - | 3 | 229 | |||
AY010898.1 | - | 1 | 76 | |||
BC018443.1 | - | 2 | 350 | 3601865 | ||
BC032928.1 | - | 2 | 265 | 5120275 | ||
BC096496.1 | - | 3 | 741 | 30738460 | ||
BY130221.1 | - | 3 | 233 | |||
BY228140.1 | - | 2 | 340 | |||
BY721478.1 | - | 3 | 227 |
