siRNA Details
Gene Symbol: Calm4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020036.4 | NP_064420.2 | 1 | 198 | ||
GenBank mRNA | AK003648.1 | - | 1 | 199 | ||
AK009664.1 | - | 1 | 200 | |||
AK009956.1 | - | 1 | 199 | |||
AY293058.1 | - | 1 | 181 | |||
BC060284.1 | - | 1 | 184 | 30293362 | ||
BC145379.1 | - | 1 | 154 | 9053937 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
