siRNA Details
Gene Symbol: Dhx58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030150.2 | NP_084426.2 | 7 | 1015 | ||
GenBank mRNA | AF316999.1 | - | 6 | 914 | ||
AK153342.1 | - | 7 | 944 | |||
BC029209.1 | - | 6 | 925 | 2651254 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
