siRNA Details
Gene Symbol: Hspb8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF250139.1 | - | 1 | 370 | ||
AF273453.1 | - | 1 | 716 | |||
AK005052.1 | - | 1 | 738 | |||
BC003971.1 | - | 1 | 726 | |||
BC011219.1 | - | 1 | 719 | 4195285 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
